Product Details

SNP ID
rs113344381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:69082638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTGTAATTTCGTGTACGTTAAA[A/T]ATATGTGATCAAGTGAAAAGAGACT
Phenotype
MIM: 616492
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EWSAT1 PubMed Links

Gene Details

Gene
EWSAT1
Gene Name
Ewing sarcoma associated transcript 1
There are no transcripts associated with this gene.

View Full Product Details