Product Details

SNP ID
rs114345862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43203223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGAGTCTAAGGAAGGTGTTCTC[A/G]GGTGGGTTTCGCTCAAAATTGGAGA
Phenotype
MIM: 607089 MIM: 177070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNDBP1 PubMed Links

Gene Details

Gene
CCNDBP1
Gene Name
cyclin D1 binding protein 1
There are no transcripts associated with this gene.

Gene
EPB42
Gene Name
erythrocyte membrane protein band 4.2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000119.2 1703 Silent Mutation NP_000110.2
NM_001114134.1 1703 Silent Mutation NP_001107606.1
XM_005254225.1 1703 Silent Mutation XP_005254282.1
XM_011521349.2 1703 Silent Mutation XP_011519651.1
XM_011521350.2 1703 Silent Mutation XP_011519652.1
XM_011521351.2 1703 Silent Mutation XP_011519653.1
XM_011521352.2 1703 Silent Mutation XP_011519654.1
XM_011521353.2 1703 Silent Mutation XP_011519655.1
XM_011521354.2 1703 Silent Mutation XP_011519656.1

View Full Product Details