Product Details

SNP ID
rs117641963
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34083817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAAATTGAAACTACATCAGCAA[C/T]ATATGGCTCAGATTATCAGAGAATT
Phenotype
MIM: 605265
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AVEN PubMed Links

Gene Details

Gene
AVEN
Gene Name
apoptosis and caspase activation inhibitor
There are no transcripts associated with this gene.

Gene
EMC7
Gene Name
ER membrane protein complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020154.2 Intron NP_064539.1

View Full Product Details