Product Details
- SNP ID
-
rs117764534
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:56631916 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTTCCTGACAAACGTGTTGTATT[G/T]GTTTTTTCTTTTCCACTTTATCTGC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF280D
PubMed Links
Gene Details
- Gene
- ZNF280D
- Gene Name
- zinc finger protein 280D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001002843.2 |
2381 |
Missense Mutation |
CAA,CCA |
Q828P |
NP_001002843.1 |
NM_001002844.2 |
2381 |
Intron |
|
|
NP_001002844.1 |
NM_001288588.1 |
2381 |
Missense Mutation |
CAA,CCA |
Q841P |
NP_001275517.1 |
NM_001288589.1 |
2381 |
Intron |
|
|
NP_001275518.1 |
NM_017661.3 |
2381 |
Missense Mutation |
CAA,CCA |
Q841P |
NP_060131.2 |
XM_005254481.3 |
2381 |
Intron |
|
|
XP_005254538.2 |
XM_005254483.3 |
2381 |
Intron |
|
|
XP_005254540.2 |
XM_005254484.3 |
2381 |
Intron |
|
|
XP_005254541.2 |
XM_005254485.3 |
2381 |
Intron |
|
|
XP_005254542.2 |
XM_011521701.1 |
2381 |
Missense Mutation |
CAA,CCA |
Q776P |
XP_011520003.1 |
XM_011521702.1 |
2381 |
Missense Mutation |
CAA,CCA |
Q841P |
XP_011520004.1 |
XM_011521704.2 |
2381 |
Intron |
|
|
XP_011520006.1 |
XM_011521707.2 |
2381 |
Intron |
|
|
XP_011520009.1 |
XM_011521708.1 |
2381 |
Missense Mutation |
CAA,CCA |
Q735P |
XP_011520010.1 |
XM_011521709.1 |
2381 |
Missense Mutation |
CAA,CCA |
Q735P |
XP_011520011.1 |
XM_011521710.1 |
2381 |
Missense Mutation |
CAA,CCA |
Q735P |
XP_011520012.1 |
XM_011521711.1 |
2381 |
Intron |
|
|
XP_011520013.1 |
XM_017022344.1 |
2381 |
Missense Mutation |
CAA,CCA |
Q841P |
XP_016877833.1 |
XM_017022345.1 |
2381 |
Missense Mutation |
CAA,CCA |
Q841P |
XP_016877834.1 |
XM_017022346.1 |
2381 |
Intron |
|
|
XP_016877835.1 |
XM_017022347.1 |
2381 |
Intron |
|
|
XP_016877836.1 |
XM_017022348.1 |
2381 |
Intron |
|
|
XP_016877837.1 |
XM_017022349.1 |
2381 |
Missense Mutation |
CAA,CCA |
Q353P |
XP_016877838.1 |
View Full Product Details