Product Details

SNP ID
rs118142218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74541747 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGGGCTTACAGGGGCCCGGGG[C/T]CAAAAGAATCCGGCAGCAAAAGCAG
Phenotype
MIM: 612457
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARID3B PubMed Links

Gene Details

Gene
ARID3B
Gene Name
AT-rich interaction domain 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307939.1 Intron NP_001294868.1
NM_006465.3 Intron NP_006456.1

View Full Product Details