Product Details

SNP ID
rs138384328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75598636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGCCCACCAAGGGAGTGCTTC[C/T]GGGGCTGTAGTGGGTCTGTTTGTGG
Phenotype
MIM: 607902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNUPN PubMed Links

Gene Details

Gene
SNUPN
Gene Name
snurportin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042581.1 950 Missense Mutation AGA,GGA R269G NP_001036046.1
NM_001042588.1 950 Missense Mutation AGA,GGA R269G NP_001036053.1
NM_005701.3 950 Missense Mutation AGA,GGA R269G NP_005692.1

View Full Product Details