Product Details

SNP ID
rs139049909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78288346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGTGACAAGGAGCTGGGAGTCC[C/G]GGGAAAAGGTCAAGGAGCGAATGGG
Phenotype
MIM: 609540
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAJA4 PubMed Links

Gene Details

Gene
DNAJA4
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
There are no transcripts associated with this gene.

Gene
WDR61
Gene Name
WD repeat domain 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303247.1 803 Missense Mutation CCG,CGG P200R NP_001290176.1
NM_001303248.1 803 Missense Mutation CCG,CGG P107R NP_001290177.1
NM_025234.2 803 Missense Mutation CCG,CGG P200R NP_079510.1
XM_011522094.1 803 Missense Mutation CCG,CGG P200R XP_011520396.1
XM_017022637.1 803 Missense Mutation CCG,CGG P200R XP_016878126.1

View Full Product Details