Product Details

SNP ID
rs139346626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40335195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCGGGCCAGGGCGCCCTGTCCTG[C/T]GCCTCACACCTGCTGTGCCTCTCGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C15orf52 PubMed Links

Gene Details

Gene
C15orf52
Gene Name
chromosome 15 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207380.2 1584 Missense Mutation CAC,CGC H523R NP_997263.2
Gene
INAFM2
Gene Name
InaF motif containing 2
There are no transcripts associated with this gene.

View Full Product Details