Product Details

SNP ID
rs139583673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63907241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAATTATATTGCATCGGGGCCC[A/G]TATCAGTGAGCTCATTTTAATTTAA
Phenotype
MIM: 616567
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAPK2 PubMed Links

Gene Details

Gene
DAPK2
Gene Name
death associated protein kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014326.3 2423 UTR 3 NP_055141.2
XM_011521413.2 2423 Intron XP_011519715.1
XM_011521414.1 2423 Intron XP_011519716.1
XM_011521415.2 2423 Intron XP_011519717.1
XM_011521416.2 2423 Intron XP_011519718.1
XM_011521417.1 2423 Intron XP_011519719.1
XM_011521421.2 2423 Intron XP_011519723.1
XM_017022043.1 2423 Intron XP_016877532.1
XM_017022044.1 2423 Intron XP_016877533.1
XM_017022045.1 2423 Intron XP_016877534.1
XM_017022046.1 2423 Intron XP_016877535.1
XM_017022047.1 2423 Intron XP_016877536.1
XM_017022048.1 2423 Intron XP_016877537.1
XM_017022049.1 2423 Intron XP_016877538.1
XM_017022050.1 2423 Intron XP_016877539.1
XM_017022051.1 2423 Intron XP_016877540.1
XM_017022052.1 2423 Intron XP_016877541.1

View Full Product Details