Product Details

SNP ID
rs141408568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90241012 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGACTTTGTTTATGGGCAGAAGG[A/C]TCTCATGGCAGAAGGGATTCAGTGG
Phenotype
MIM: 602293
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CIB1 PubMed Links

Gene Details

Gene
CIB1
Gene Name
calcium and integrin binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277764.1 283 Intron NP_001264693.1
NM_006384.3 283 Intron NP_006375.2
XM_006720375.1 283 Intron XP_006720438.1
Gene
GDPGP1
Gene Name
GDP-D-glucose phosphorylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013657.2 283 Missense Mutation GAT,GCT D35A NP_001013679.2
NM_001322811.1 283 Missense Mutation GAT,GCT D35A NP_001309740.1
Gene
TTLL13P
Gene Name
tubulin tyrosine ligase like 13, pseudogene
There are no transcripts associated with this gene.

View Full Product Details