Product Details
- SNP ID
-
rs141577275
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:64968538 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCGTTGATTATCAGAGGCCGGGG[A/G]AAGAAAGGGAACAGGAGTTATTGTT
- Phenotype
-
MIM: 608181
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANKDD1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112449052] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANKDD1A
- Gene Name
- ankyrin repeat and death domain containing 1A
There are no transcripts associated with this gene.
- Gene
- SPG21
- Gene Name
- spastic paraplegia 21 (autosomal recessive, Mast syndrome)
View Full Product Details