Product Details

SNP ID
rs141659576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:82271801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGGACACTGTCCCAACCTATAA[A/G]GGTAACTTGCTGTTTCATACATGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFL1 PubMed Links

Gene Details

Gene
EFL1
Gene Name
elongation factor like GTPase 1
There are no transcripts associated with this gene.

Gene
SAXO2
Gene Name
stabilizer of axonemal microtubules 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008226.1 Intron NP_001008227.1

View Full Product Details