Product Details

SNP ID
rs141944790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63597455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGGAGTGCCTGCTGCACCTCTT[C/T]TCCTTCCTAGACAAGGACAGCAGGA
Phenotype
MIM: 609088 MIM: 604728
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL22 PubMed Links

Gene Details

Gene
FBXL22
Gene Name
F-box and leucine rich repeat protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203373.2 65 Silent Mutation TTC,TTT F21F NP_976307.2
XM_005254320.3 65 Silent Mutation TTC,TTT F21F XP_005254377.1
XM_006720478.1 65 Intron XP_006720541.1
XM_011521468.2 65 UTR 5 XP_011519770.1
XM_011521469.1 65 UTR 5 XP_011519771.1
XM_011521472.2 65 UTR 5 XP_011519774.1
XM_017022086.1 65 Silent Mutation TTC,TTT F21F XP_016877575.1
Gene
USP3
Gene Name
ubiquitin specific peptidase 3
There are no transcripts associated with this gene.

Gene
USP3-AS1
Gene Name
USP3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details