Product Details

SNP ID
rs143063914
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40363904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTTCTCCAGCCCCCTCACAGCG[C/T]GATGGGACCTGGAAGCCACCCGCTG
Phenotype
MIM: 607503
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DISP2 PubMed Links
Additional Information
For this assay, SNP(s) [rs12443160] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DISP2
Gene Name
dispatched RND transporter family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033510.2 2657 Silent Mutation CGC,CGT R133R NP_277045.1
XM_011522125.2 2657 Silent Mutation CGC,CGT R133R XP_011520427.1
XM_017022687.1 2657 Silent Mutation CGC,CGT R133R XP_016878176.1
Gene
PHGR1
Gene Name
proline, histidine and glycine rich 1
There are no transcripts associated with this gene.

View Full Product Details