Product Details

SNP ID
rs143234818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89835617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGTTTTTCACAGCAGACACAGC[C/G]CGCGCAGGGGCTGCTGAAAGGCCAC
Phenotype
MIM: 602416
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AP3S2 PubMed Links

Gene Details

Gene
AP3S2
Gene Name
adaptor related protein complex 3 sigma 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005829.4 916 Silent Mutation CGC,CGG R160R NP_005820.1
Gene
C15orf38-AP3S2
Gene Name
C15orf38-AP3S2 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199058.1 916 Silent Mutation CGC,CGG R361R NP_001185987.1

View Full Product Details