Product Details

SNP ID
rs143445521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42142075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCACGTTGTTCAGGACGTTGTAT[C/T]GACTGAGGGCCACCAGCCGATAAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G4F PubMed Links

Gene Details

Gene
PLA2G4F
Gene Name
phospholipase A2 group IVF
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213600.3 2551 Missense Mutation CAA,CGA Q820R NP_998765.3

View Full Product Details