Product Details

SNP ID
rs144563180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72162337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGCTGCTCTAGCCGAGAAATAC[G/T]GAACGCCAGGTAGGATGAGGACATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GRAMD2 PubMed Links

Gene Details

Gene
GRAMD2
Gene Name
GRAM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012642.2 1205 Missense Mutation NP_001012660.1
XM_011521327.2 1205 Missense Mutation XP_011519629.1
XM_011521328.2 1205 Missense Mutation XP_011519630.1
XM_011521329.2 1205 Missense Mutation XP_011519631.1
XM_011521330.2 1205 Missense Mutation XP_011519632.1
XM_011521331.2 1205 Missense Mutation XP_011519633.1
XM_011521332.2 1205 Missense Mutation XP_011519634.1
XM_011521333.2 1205 Missense Mutation XP_011519635.1
XM_017021997.1 1205 Missense Mutation XP_016877486.1
XM_017021998.1 1205 Missense Mutation XP_016877487.1

View Full Product Details