Product Details

SNP ID
rs144666544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75605217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTTTGAATGCATCCAGTAGAAT[C/T]GGAAATCAGTCTGCCACAGAGAAGG
Phenotype
MIM: 607902
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNUPN PubMed Links

Gene Details

Gene
SNUPN
Gene Name
snurportin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042581.1 756 Missense Mutation CAA,CGA Q204R NP_001036046.1
NM_001042588.1 756 Missense Mutation CAA,CGA Q204R NP_001036053.1
NM_005701.3 756 Missense Mutation CAA,CGA Q204R NP_005692.1

View Full Product Details