Product Details

SNP ID
rs144969865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75674771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCCTAACCTGCTCCAAAGCCAC[C/T]GACACATGCATGTTTAGCCCTTGGC
Phenotype
MIM: 601172
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSPG4 PubMed Links

Gene Details

Gene
CSPG4
Gene Name
chondroitin sulfate proteoglycan 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001897.4 7841 UTR 3 NP_001888.2

View Full Product Details