Product Details

SNP ID
rs145226318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:30905124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTGAAAGTCATTTGTTTGGGAA[A/G]CCTAGCATCTAAATTGTCCAGAAAA
Phenotype
MIM: 613534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAN1 PubMed Links

Gene Details

Gene
FAN1
Gene Name
FANCD2/FANCI-associated nuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146094.1 756 Missense Mutation AAC,AGC N154S NP_001139566.1
NM_001146095.1 756 Missense Mutation AAC,AGC N154S NP_001139567.1
NM_001146096.1 756 Missense Mutation AAC,AGC N154S NP_001139568.1
NM_014967.4 756 Missense Mutation AAC,AGC N154S NP_055782.3
XM_005254232.4 756 Missense Mutation AAC,AGC N154S XP_005254289.1
XM_005254234.4 756 Missense Mutation AAC,AGC N154S XP_005254291.1
XM_005254235.3 756 Missense Mutation AAC,AGC N154S XP_005254292.1
XM_011521370.2 756 Intron XP_011519672.1
XM_011521371.1 756 UTR 5 XP_011519673.1
XM_011521372.2 756 Missense Mutation AAC,AGC N154S XP_011519674.1
XM_017022012.1 756 UTR 5 XP_016877501.1
XM_017022013.1 756 UTR 5 XP_016877502.1

View Full Product Details