Product Details
- SNP ID
-
rs145448730
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:29269088 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAATGCTTCTTGGTGGGGTAGAC[A/C]CCTAAGCGCCGCAGAAAGTCCCAGG
- Phenotype
-
MIM: 608243
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM189A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61749506] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM189A1
- Gene Name
- family with sequence similarity 189 member A1
- Gene
- NSMCE3
- Gene Name
- NSE3 homolog, SMC5-SMC6 complex component
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138704.3 |
729 |
Silent Mutation |
GGG,GGT |
G206G |
NP_619649.1 |
View Full Product Details