Product Details
- SNP ID
-
rs145719547
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:49331818 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACTTCTCAAAGTCCTGTTTCACTT[C/T]ATGAGGTTTCTTGGTAGCCTTTGCT
- Phenotype
-
MIM: 137028
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FAM227B
PubMed Links
Gene Details
- Gene
- FAM227B
- Gene Name
- family with sequence similarity 227 member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152647.2 |
1725 |
Missense Mutation |
AAA,GAA |
K461E |
NP_689860.2 |
XM_005254213.3 |
1725 |
Missense Mutation |
AAA,GAA |
K486E |
XP_005254270.1 |
XM_005254214.3 |
1725 |
Missense Mutation |
AAA,GAA |
K486E |
XP_005254271.1 |
XM_005254215.3 |
1725 |
Missense Mutation |
AAA,GAA |
K474E |
XP_005254272.1 |
XM_005254216.3 |
1725 |
Missense Mutation |
AAA,GAA |
K452E |
XP_005254273.1 |
XM_005254219.3 |
1725 |
Silent Mutation |
TAA,TGA |
*381* |
XP_005254276.1 |
XM_005254220.3 |
1725 |
Intron |
|
|
XP_005254277.1 |
XM_006720423.2 |
1725 |
Missense Mutation |
AAA,GAA |
K486E |
XP_006720486.1 |
XM_006720424.2 |
1725 |
Missense Mutation |
AAA,GAA |
K461E |
XP_006720487.1 |
XM_006720426.2 |
1725 |
Missense Mutation |
AAA,GAA |
K439E |
XP_006720489.1 |
XM_006720427.3 |
1725 |
Missense Mutation |
AAA,GAA |
K403E |
XP_006720490.1 |
XM_011521319.2 |
1725 |
Missense Mutation |
AAA,GAA |
K486E |
XP_011519621.1 |
XM_011521320.1 |
1725 |
Missense Mutation |
AAA,GAA |
K352E |
XP_011519622.1 |
XM_011521321.2 |
1725 |
Intron |
|
|
XP_011519623.1 |
XM_011521322.1 |
1725 |
Missense Mutation |
AAA,GAA |
K340E |
XP_011519624.1 |
XM_011521324.1 |
1725 |
Intron |
|
|
XP_011519626.1 |
XM_011521325.2 |
1725 |
Intron |
|
|
XP_011519627.1 |
XM_011521326.1 |
1725 |
Intron |
|
|
XP_011519628.1 |
XM_017021990.1 |
1725 |
Missense Mutation |
AAA,GAA |
K405E |
XP_016877479.1 |
XM_017021991.1 |
1725 |
Missense Mutation |
AAA,GAA |
K403E |
XP_016877480.1 |
XM_017021992.1 |
1725 |
Missense Mutation |
AAA,GAA |
K386E |
XP_016877481.1 |
XM_017021993.1 |
1725 |
Missense Mutation |
AAA,GAA |
K380E |
XP_016877482.1 |
XM_017021994.1 |
1725 |
Missense Mutation |
AAA,GAA |
K372E |
XP_016877483.1 |
XM_017021995.1 |
1725 |
Missense Mutation |
ATA,ATG |
I399M |
XP_016877484.1 |
XM_017021996.1 |
1725 |
Missense Mutation |
AAA,GAA |
K352E |
XP_016877485.1 |
- Gene
- GALK2
- Gene Name
- galactokinase 2
View Full Product Details