Product Details

SNP ID
rs147061047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74133008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACGTCGCTGTGGCTGGCGCACA[A/G]TGAGGTGCGCACCGTGGAGCCAGGC
Phenotype
MIM: 614179
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISLR2 PubMed Links

Gene Details

Gene
ISLR2
Gene Name
immunoglobulin superfamily containing leucine rich repeat 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130136.1 862 Missense Mutation AAT,AGT N85S NP_001123608.1
NM_001130137.1 862 Missense Mutation AAT,AGT N85S NP_001123609.1
NM_001130138.1 862 Missense Mutation AAT,AGT N85S NP_001123610.1
NM_020851.2 862 Missense Mutation AAT,AGT N85S NP_065902.1
XM_011521840.2 862 Missense Mutation AAT,AGT N85S XP_011520142.1
XM_011521841.1 862 Missense Mutation AAT,AGT N85S XP_011520143.1
XM_017022446.1 862 Missense Mutation AAT,AGT N85S XP_016877935.1
Gene
LOC283731
Gene Name
uncharacterized LOC283731
There are no transcripts associated with this gene.

View Full Product Details