Product Details

SNP ID
rs148118120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75675721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCTCGGTGTCACCAGCCAGGCC[A/G]TTGCGGGGCTTGGCAGTCAGGACCT
Phenotype
MIM: 601172
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CSPG4 PubMed Links

Gene Details

Gene
CSPG4
Gene Name
chondroitin sulfate proteoglycan 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001897.4 6891 Silent Mutation AAC,AAT N2266N NP_001888.2

View Full Product Details