Product Details

SNP ID
rs148453374
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63597738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCAATGATTTCCTCCTCCGGGTG[A/T]GCGACAGGTAGGCCACCTTGCCTCC
Phenotype
MIM: 609088 MIM: 604728
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
FBXL22 PubMed Links
Additional Information
For this assay, SNP(s) [rs8035931] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FBXL22
Gene Name
F-box and leucine rich repeat protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203373.2 348 Missense Mutation AGC,TGC S116C NP_976307.2
XM_005254320.3 348 Missense Mutation AGC,TGC S116C XP_005254377.1
XM_006720478.1 348 UTR 5 XP_006720541.1
XM_011521468.2 348 UTR 5 XP_011519770.1
XM_011521469.1 348 UTR 5 XP_011519771.1
XM_011521472.2 348 UTR 5 XP_011519774.1
XM_017022086.1 348 Missense Mutation AGC,TGC S116C XP_016877575.1
Gene
USP3
Gene Name
ubiquitin specific peptidase 3
There are no transcripts associated with this gene.

Gene
USP3-AS1
Gene Name
USP3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details