Product Details

SNP ID
rs149044800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75206429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACTCCTACCCAGGCCCACCACT[G/T]GCAGCACCCAAACCTGTCTACCGCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C15orf39 PubMed Links

Gene Details

Gene
C15orf39
Gene Name
chromosome 15 open reading frame 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015492.4 611 Silent Mutation CTG,CTT L127L NP_056307.2

View Full Product Details