Product Details

SNP ID
rs149818706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42162096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGAACCGTTTCTTTTGTGCATTT[C/T]CTTCCAAGACCTTTTCCAGGAAGAT
Phenotype
MIM: 612188
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G4F PubMed Links

Gene Details

Gene
PLA2G4F
Gene Name
phospholipase A2 group IVF
There are no transcripts associated with this gene.

Gene
VPS39
Gene Name
VPS39, HOPS complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301138.1 2574 Missense Mutation GAA,GGA E810G NP_001288067.1
NM_015289.3 2574 Missense Mutation GAA,GGA E799G NP_056104.2
XM_011521403.2 2574 Missense Mutation GAA,GGA E810G XP_011519705.1
XM_011521404.2 2574 Missense Mutation GAA,GGA E799G XP_011519706.1
XM_017022036.1 2574 Missense Mutation GAA,GGA E810G XP_016877525.1
XM_017022037.1 2574 Missense Mutation GAA,GGA E799G XP_016877526.1
XM_017022038.1 2574 Missense Mutation GAA,GGA E675G XP_016877527.1
XM_017022039.1 2574 Missense Mutation GAA,GGA E442G XP_016877528.1

View Full Product Details