Product Details

SNP ID
rs150503184
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75375738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGATCACATACACCATCTTGTAG[G/T]AATTCAGCTTGAATCTACACTCCAG
Phenotype
MIM: 154580 MIM: 607776
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MAN2C1 PubMed Links

Gene Details

Gene
MAN2C1
Gene Name
mannosidase alpha class 2C member 1
There are no transcripts associated with this gene.

Gene
SIN3A
Gene Name
SIN3 transcription regulator family member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145357.1 3700 Missense Mutation TAC,TCC Y1173S NP_001138829.1
NM_001145358.1 3700 Missense Mutation TAC,TCC Y1173S NP_001138830.1
NM_015477.2 3700 Missense Mutation TAC,TCC Y1173S NP_056292.1
XM_006720465.3 3700 Missense Mutation TAC,TCC Y1173S XP_006720528.1
XM_006720466.3 3700 Missense Mutation TAC,TCC Y1173S XP_006720529.1
XM_006720467.3 3700 Missense Mutation TAC,TCC Y1173S XP_006720530.1

View Full Product Details