Product Details

SNP ID
rs150766109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75675773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGTCATGCTTGCCCGTCTTGTTG[C/G]GTTTTCGGAGGTAGAAGAGCAGGGG
Phenotype
MIM: 601172
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CSPG4 PubMed Links

Gene Details

Gene
CSPG4
Gene Name
chondroitin sulfate proteoglycan 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001897.4 6839 Missense Mutation CCC,CGC P2249R NP_001888.2

View Full Product Details