Product Details
- SNP ID
-
rs150933904
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:38696612 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACAGGTTTTCTGTTCTGATATGG[A/G]GCTACAAGGGGCCCAAGAGGACCTG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C15orf53
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7165988] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C15orf53
- Gene Name
- chromosome 15 open reading frame 53
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207444.2 |
15 |
Missense Mutation |
GAG,GGG |
E2G |
NP_997327.1 |
View Full Product Details