Product Details

SNP ID
rs138046858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30992550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACACATCACACGCACACGCAC[A/G]CTGGGGTGTCCACACGTCTCGAGCA
Phenotype
MIM: 607764 MIM: 611052 MIM: 601485
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSD3B7 PubMed Links

Gene Details

Gene
HSD3B7
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
There are no transcripts associated with this gene.

Gene
SETD1A
Gene Name
SET domain containing 1A
There are no transcripts associated with this gene.

Gene
STX1B
Gene Name
syntaxin 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052874.4 1380 UTR 3 NP_443106.1
XM_017022893.1 1380 UTR 3 XP_016878382.1

View Full Product Details