Product Details

SNP ID
rs138180373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28593278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTCATGTCTTCATAGAAGAGGT[A/G]GAGAACAGGGTGGGTGCGGCTCAGC
Phenotype
MIM: 613374 MIM: 601292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SGF29 PubMed Links

Gene Details

Gene
SGF29
Gene Name
SAGA complex associated factor 29
There are no transcripts associated with this gene.

Gene
SULT1A2
Gene Name
sulfotransferase family 1A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001054.3 785 Missense Mutation CAC,TAC H190Y NP_001045.1
NM_177528.2 785 Missense Mutation CAC,TAC H190Y NP_803564.1
XM_006721077.3 785 Intron XP_006721140.1
XM_017023600.1 785 Missense Mutation CAC,TAC H190Y XP_016879089.1
XM_017023601.1 785 Missense Mutation CAC,TAC H190Y XP_016879090.1
XM_017023602.1 785 Missense Mutation CAC,TAC H190Y XP_016879091.1
XM_017023603.1 785 Intron XP_016879092.1

View Full Product Details