Product Details

SNP ID
rs139042144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75478648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACAACTGTGGCCTCCACTAATTT[C/T]GGGGGTGCGAGGCGGTGGATGATGC
Phenotype
MIM: 605294
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST6 PubMed Links

Gene Details

Gene
CHST6
Gene Name
carbohydrate sulfotransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021615.4 1976 Missense Mutation CCA,CTA P394L NP_067628.1
XM_005255955.4 1976 Missense Mutation XP_005256012.1
XM_011523085.2 1976 Missense Mutation XP_011521387.1

View Full Product Details