Product Details

SNP ID
rs139174776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2166296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTCAGGCTCCTGGCCAGCCTCC[A/G]AAAGGTCCCGTGTGCCCGGAGGTCT
Phenotype
MIM: 606692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAF7 PubMed Links

Gene Details

Gene
TRAF7
Gene Name
TNF receptor associated factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032271.2 Intron NP_115647.2
XM_005255627.4 Intron XP_005255684.1
XM_011522700.1 Intron XP_011521002.1

View Full Product Details