Product Details

SNP ID
rs139832768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70350568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTCTATTCAGATGACCAATTT[G/T]CAGATCAAGGAAGAGAAAGTCAAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DDX19A PubMed Links

Gene Details

Gene
DDX19A
Gene Name
DEAD-box helicase 19A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320522.1 232 Missense Mutation TTG,TTT L23F NP_001307451.1
NM_001320525.1 232 UTR 5 NP_001307454.1
NM_001320526.1 232 UTR 5 NP_001307455.1
NM_001320527.1 232 UTR 5 NP_001307456.1
NM_018332.4 232 Missense Mutation TTG,TTT L23F NP_060802.1
Gene
LOC100506083
Gene Name
uncharacterized LOC100506083
There are no transcripts associated with this gene.

View Full Product Details