Product Details

SNP ID
rs140310025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:86578845 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCGCGGCTGCTCTAGCTGCCT[C/T]GGGCCGGGCCGAGACCCCGCAGAAG
Phenotype
MIM: 602402 MIM: 603252
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXC2 PubMed Links

Gene Details

Gene
FOXC2
Gene Name
forkhead box C2
There are no transcripts associated with this gene.

Gene
FOXL1
Gene Name
forkhead box L1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005250.2 337 Missense Mutation TCG,TTG S41L NP_005241.1

View Full Product Details