Product Details

SNP ID
rs140374059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:53600566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACATCAACAAATTAATATTGTA[C/T]GGATGTCATTTCAAAGCCTTCTAAA
Phenotype
MIM: 610937
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPGRIP1L PubMed Links

Gene Details

Gene
RPGRIP1L
Gene Name
RPGRIP1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005255867.1 5502 UTR 3 XP_005255924.1
XM_005255868.1 5502 UTR 3 XP_005255925.1
XM_011522970.1 5502 Intron XP_011521272.1
XM_011522971.2 5502 Intron XP_011521273.1
XM_011522973.2 5502 Intron XP_011521275.1
XM_017023094.1 5502 UTR 3 XP_016878583.1
XM_017023095.1 5502 UTR 3 XP_016878584.1
XM_017023096.1 5502 Intron XP_016878585.1
XM_017023097.1 5502 Intron XP_016878586.1
XM_017023098.1 5502 UTR 3 XP_016878587.1
XM_017023099.1 5502 UTR 3 XP_016878588.1
XM_017023100.1 5502 Intron XP_016878589.1

View Full Product Details