Product Details

SNP ID
rs140654547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:55325114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATGTCCTTCCCACACTTTGGAC[A/G]CCCGTACCGCGGCGCTTCCCAGGTA
Phenotype
MIM: 606196
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRX6 PubMed Links

Gene Details

Gene
IRX6
Gene Name
iroquois homeobox 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024335.2 556 Missense Mutation CAC,CGC H8R NP_077311.2
XM_005256137.2 556 Missense Mutation CAC,CGC H8R XP_005256194.1

View Full Product Details