Product Details

SNP ID
rs141121785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56963018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCCCTCCCCTCTAGTGAACCAC[A/G]AGACTGCCAAGGTGATCCAGACCGC
Phenotype
MIM: 118470
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CETP PubMed Links

Gene Details

Gene
CETP
Gene Name
cholesteryl ester transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000078.2 144 Missense Mutation AAG,GAG K43E NP_000069.2
NM_001286085.1 144 Missense Mutation AAG,GAG K43E NP_001273014.1
XM_006721124.3 144 Missense Mutation AAG,GAG K43E XP_006721187.1

View Full Product Details