Product Details

SNP ID
rs141262832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3402079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCAAGAGAGACACATAATAGC[C/T]AAACTAGAAGAGAAACGGGGCCCTC
Phenotype
MIM: 603900
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF174 PubMed Links

Gene Details

Gene
ZNF174
Gene Name
zinc finger protein 174
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032292.2 890 Silent Mutation GCC,GCT A25A NP_001027463.1
NM_003450.2 890 Silent Mutation GCC,GCT A25A NP_003441.1
Gene
ZSCAN32
Gene Name
zinc finger and SCAN domain containing 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284527.1 890 Intron NP_001271456.1
NM_001284528.1 890 Intron NP_001271457.1
NM_001284529.1 890 Intron NP_001271458.1
NM_001324340.1 890 Intron NP_001311269.1
NM_001324341.1 890 Intron NP_001311270.1
NM_001324342.1 890 Intron NP_001311271.1
NM_001324343.1 890 Intron NP_001311272.1
NM_001324344.1 890 Intron NP_001311273.1
NM_001324345.1 890 Intron NP_001311274.1
NM_001324346.1 890 Intron NP_001311275.1
NM_017810.3 890 Intron NP_060280.2

View Full Product Details