Product Details

SNP ID
rs141266321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56625871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGAAATGGACCCCAACTGCTCTT[A/G]CGCCACTGGTAAGGGAAGCTCTGCG
Phenotype
MIM: 156351 MIM: 156356 MIM: 156358 MIM: 156357
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MT1E PubMed Links

Gene Details

Gene
MT1E
Gene Name
metallothionein 1E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175617.3 199 Missense Mutation TAC,TGC Y7C NP_783316.2
XM_005255956.4 199 Missense Mutation TAC,TGC Y7C XP_005256013.1
Gene
MT1JP
Gene Name
metallothionein 1J, pseudogene
There are no transcripts associated with this gene.

Gene
MT1L
Gene Name
metallothionein 1L (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
MT1M
Gene Name
metallothionein 1M
There are no transcripts associated with this gene.

View Full Product Details