Product Details

SNP ID
rs141440927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4426336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAACGCCCCGTAGATCGTGAACC[C/T]TCACCTCTCTGTCAACATTTCTGGA
Phenotype
MIM: 611668 MIM: 608382
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CORO7 PubMed Links

Gene Details

Gene
CORO7
Gene Name
coronin 7
There are no transcripts associated with this gene.

Gene
CORO7-PAM16
Gene Name
CORO7-PAM16 readthrough
There are no transcripts associated with this gene.

Gene
DNAJA3
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135110.2 Intron NP_001128582.1
NM_001286516.1 Intron NP_001273445.1
NM_005147.5 Intron NP_005138.3

View Full Product Details