Product Details
- SNP ID
-
rs141657089
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:3383146 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATGCACTGGTAAGGCTTTTCCCC[A/G]GTATGGGTCCTCTGGTGGACAATGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZSCAN32
PubMed Links
Gene Details
- Gene
- ZSCAN32
- Gene Name
- zinc finger and SCAN domain containing 32
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001284527.1 |
2063 |
Silent Mutation |
ACC,ACT |
T600T |
NP_001271456.1 |
NM_001284528.1 |
2063 |
Silent Mutation |
ACC,ACT |
T311T |
NP_001271457.1 |
NM_001284529.1 |
2063 |
Silent Mutation |
ACC,ACT |
T311T |
NP_001271458.1 |
NM_001324340.1 |
2063 |
Silent Mutation |
ACC,ACT |
T388T |
NP_001311269.1 |
NM_001324341.1 |
2063 |
Silent Mutation |
ACC,ACT |
T388T |
NP_001311270.1 |
NM_001324342.1 |
2063 |
Silent Mutation |
ACC,ACT |
T388T |
NP_001311271.1 |
NM_001324343.1 |
2063 |
Silent Mutation |
ACC,ACT |
T467T |
NP_001311272.1 |
NM_001324344.1 |
2063 |
Silent Mutation |
ACC,ACT |
T388T |
NP_001311273.1 |
NM_001324345.1 |
2063 |
Silent Mutation |
ACC,ACT |
T388T |
NP_001311274.1 |
NM_001324346.1 |
2063 |
Silent Mutation |
ACC,ACT |
T527T |
NP_001311275.1 |
NM_017810.3 |
2063 |
Silent Mutation |
ACC,ACT |
T388T |
NP_060280.2 |
XM_005255403.2 |
2063 |
Silent Mutation |
ACC,ACT |
T513T |
XP_005255460.1 |
XM_011522555.2 |
2063 |
Silent Mutation |
ACC,ACT |
T600T |
XP_011520857.1 |
XM_017023371.1 |
2063 |
Silent Mutation |
ACC,ACT |
T600T |
XP_016878860.1 |
XM_017023372.1 |
2063 |
Silent Mutation |
ACC,ACT |
T513T |
XP_016878861.1 |
XM_017023373.1 |
2063 |
Silent Mutation |
ACC,ACT |
T513T |
XP_016878862.1 |
View Full Product Details