Product Details

SNP ID
rs142542904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1358336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTGCTGTCTCTGAACTTGGCT[A/G]TTCATAGACGTCACATAGGTGACTC
Phenotype
MIM: 604009 MIM: 607838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BAIAP3 PubMed Links

Gene Details

Gene
BAIAP3
Gene Name
BAI1 associated protein 3
There are no transcripts associated with this gene.

Gene
GNPTG
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032520.4 Intron NP_115909.1
XM_017023782.1 Intron XP_016879271.1
XM_017023783.1 Intron XP_016879272.1
Gene
TSR3
Gene Name
TSR3, acp transferase ribosome maturation factor
There are no transcripts associated with this gene.

Gene
UNKL
Gene Name
unkempt family like zinc finger
There are no transcripts associated with this gene.

View Full Product Details