Product Details

SNP ID
rs142843064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89198697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAGGAACTGTCCCCGGGGCCCA[C/T]GCCACTGGCTTCTGCAAAGCGCCAC
Phenotype
MIM: 114019
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH15 PubMed Links

Gene Details

Gene
CDH15
Gene Name
cadherin 15
There are no transcripts associated with this gene.

Gene
SLC22A31
Gene Name
solute carrier family 22 member 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242757.1 2232 Missense Mutation ATG,GTG M77V NP_001229686.1
XM_011522897.2 2232 Missense Mutation ATG,GTG M77V XP_011521199.1
XM_011522899.2 2232 Missense Mutation ATG,GTG M77V XP_011521201.1
XM_017022962.1 2232 Missense Mutation ATG,GTG M110V XP_016878451.1
XM_017022963.1 2232 Missense Mutation ATG,GTG M84V XP_016878452.1
XM_017022964.1 2232 Missense Mutation ATG,GTG M84V XP_016878453.1
XM_017022965.1 2232 Missense Mutation ATG,GTG M84V XP_016878454.1
XM_017022966.1 2232 Missense Mutation ATG,GTG M84V XP_016878455.1
Gene
ZNF778
Gene Name
zinc finger protein 778
There are no transcripts associated with this gene.

View Full Product Details