Product Details

SNP ID
rs142894273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75293728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTCGTCATCTTCATTATCCTCT[C/T]ACAGGACCAACTTTTATTTTATTTA
Phenotype
MIM: 608108
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFDP1 PubMed Links

Gene Details

Gene
CFDP1
Gene Name
craniofacial development protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006324.2 1261 UTR 3 NP_006315.1
XM_011522814.1 1261 Intron XP_011521116.1
XM_011522815.1 1261 Intron XP_011521117.1

View Full Product Details