Product Details

SNP ID
rs143040901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84294987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGAGGAAATGCCTTTAACCGGC[A/G]TGGGGCCGGGCAGCTGCAGGAGGCA
Phenotype
MIM: 605322
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105376777 PubMed Links

Gene Details

Gene
LOC105376777
Gene Name
uncharacterized LOC105376777
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011523512.2 282 Intron XP_011521814.1
Gene
WFDC1
Gene Name
WAP four-disulfide core domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282466.1 282 Missense Mutation ATG,GTG M6V NP_001269395.1
NM_001282467.1 282 Missense Mutation ATG,GTG M6V NP_001269396.1
NM_021197.3 282 Missense Mutation ATG,GTG M6V NP_067020.2
XM_005256079.3 282 Missense Mutation ATG,GTG M6V XP_005256136.1

View Full Product Details