Product Details

SNP ID
rs143567197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31139362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCTCCCTTGAACAGCCATGCCCA[C/T]GAGGATCCAGGACCCTTCCGTCATC
Phenotype
MIM: 609912 MIM: 610560 MIM: 600823
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KAT8 PubMed Links

Gene Details

Gene
KAT8
Gene Name
lysine acetyltransferase 8
There are no transcripts associated with this gene.

Gene
PRSS36
Gene Name
protease, serine 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258290.1 2094 Missense Mutation NP_001245219.1
NM_001258291.1 2094 Missense Mutation NP_001245220.1
NM_173502.4 2094 Missense Mutation NP_775773.2
XM_011545758.2 2094 Missense Mutation XP_011544060.1
XM_017022968.1 2094 UTR 3 XP_016878457.1
XM_017022969.1 2094 Intron XP_016878458.1
XM_017022970.1 2094 UTR 3 XP_016878459.1
XM_017022971.1 2094 Missense Mutation XP_016878460.1
Gene
PRSS8
Gene Name
protease, serine 8
There are no transcripts associated with this gene.

View Full Product Details