Product Details
- SNP ID
-
rs143767765
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:86579686 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCGACCCGCATGTCCAGGGCGG[C/T]TTTTACCAGCTCGGGATCCCCTTCC
- Phenotype
-
MIM: 603252
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FOXL1
PubMed Links
Gene Details
- Gene
- FOXL1
- Gene Name
- forkhead box L1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005250.2 |
1178 |
Silent Mutation |
GGC,GGT |
G321G |
NP_005241.1 |
View Full Product Details