Product Details
- SNP ID
-
rs143791588
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31141487 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACCCACCTGAGGACACAGTGAGTG[G/T]CTGCCAGGACCCAGCCTGGGGCCAG
- Phenotype
-
MIM: 610560
MIM: 600823
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS36
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143680005] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PRSS36
- Gene Name
- protease, serine 36
- Gene
- PRSS8
- Gene Name
- protease, serine 8
There are no transcripts associated with this gene.
View Full Product Details